ENA300790810 Enamine

4-({[(4-hydroxy-3,5-dimethylphenyl)methyl]amino}methyl)-2,6-dimethylphenol

  • MDL number MFCD16817563
  • Cas Number : 47170-93-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 249...251
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66999

Safety Information

msds https://www.enaminestore.com/catalog/EN300-66999
Availability details for
Quantity
  • Availability
  • Add to Cart