ENA300790811 Enamine

1-methyl-2,4-dioxo-1H,2H,3H,4H-pyrido[2,3-d]pyrimidine-6-sulfonyl chloride

  • MDL number MFCD16817564
  • Cas Number : 1251923-69-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-67025

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67025
Availability details for
Quantity
  • Availability
  • Add to Cart