ENA300790812 Enamine

2-amino-2-(3-phenyl-1,2,4-oxadiazol-5-yl)ethan-1-ol hydrochloride

  • MDL number MFCD16817565
  • Cas Number : 1251925-45-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67027

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67027
Availability details for
Quantity
  • Availability
  • Add to Cart