ENA300790816 Enamine

1-[(2-aminophenyl)sulfanyl]-N,N-dimethylformamide hydrochloride

  • MDL number MFCD16817567
  • Cas Number : 1251923-35-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 169...171
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67063

Safety Information

msds https://www.enaminestore.com/catalog/EN300-67063
Availability details for
Quantity
  • Availability
  • Add to Cart