ENA300790828 Enamine

4-[2-(1H-imidazol-5-yl)-1,3-thiazol-4-yl]pyridine

  • MDL number MFCD16817579
  • Cas Number : 1251923-70-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67141

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67141
Availability details for
Quantity
  • Availability
  • Add to Cart