ENA300790829 Enamine

2-[4-(3-ethyl-1,2,4-oxadiazol-5-yl)phenyl]ethan-1-amine hydrochloride

  • MDL number MFCD16990637
  • Cas Number : 1251925-30-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67148

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67148
Availability details for
Quantity
  • Availability
  • Add to Cart