ENA300790834 Enamine

1-[(3-aminophenyl)sulfanyl]-N,N-dimethylformamide hydrochloride

  • MDL number MFCD16990644
  • Cas Number : 1251925-19-5
  • Molecular Weight : 232.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67186

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67186
Availability details for
Quantity
  • Availability
  • Add to Cart