ENA300790854 Enamine

2-{[(1-ethylpyrrolidin-2-yl)methyl]amino}pyridine-3-carboxylic acid

  • MDL number MFCD11523654
  • Cas Number : 1019452-29-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65514

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65514
Availability details for
Quantity
  • Availability
  • Add to Cart