ENA300790856 Enamine

methyl 2,2,2-tribromoacetate

  • MDL number MFCD00192327

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 70...74
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-20621

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-20621
pictograms GHS02,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart