ENA300790858 Enamine

4-{[4-(pyridin-2-yl)piperazin-1-yl]methyl}-1,3-thiazol-2-amine

  • MDL number MFCD12460564
  • Cas Number : 897935-20-5
  • Molecular Weight : 275.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65563

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65563
Availability details for
Quantity
  • Availability
  • Add to Cart