ENA300790860 Enamine

ethyl 5-oxo-1-phenyl-4,5-dihydro-1H-pyrazole-3-carboxylate

  • MDL number MFCD00020764
  • Cas Number : 89-33-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 184-185
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-21390

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-21390
Availability details for
Quantity
  • Availability
  • Add to Cart