ENA300790863 Enamine

(2-methanesulfonylethyl)(methyl)amine

  • MDL number MFCD02089403
  • Cas Number : 202198-18-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-65601

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65601
Availability details for
Quantity
  • Availability
  • Add to Cart