ENA300790869 Enamine

4-isothiocyanatobutanamide

  • MDL number MFCD12799362
  • Cas Number : 1100929-52-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65672

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65672
Availability details for
Quantity
  • Availability
  • Add to Cart