ENA300790873 Enamine

2-(chloromethyl)-1-ethyl-5-fluoro-1H-1,3-benzodiazole

  • MDL number MFCD12111374
  • Cas Number : 1154626-76-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65698

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65698
Availability details for
Quantity
  • Availability
  • Add to Cart