ENA300790879 Enamine

4-(3,4-dichlorophenyl)butan-2-one

  • MDL number MFCD09891026
  • Cas Number : 42160-40-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-65775

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65775
Availability details for
Quantity
  • Availability
  • Add to Cart