ENA300790898 Enamine

N-methylmethanesulfonamide

  • MDL number MFCD00776277
  • Cas Number : 1184-85-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-33617

Safety Information

msds https://www.enaminestore.com/catalog/EN300-33617
Availability details for
Quantity
  • Availability
  • Add to Cart