ENA300790902 Enamine

1-(5-bromothiophen-2-yl)-4-methylpentan-3-one

  • MDL number MFCD11655311
  • Cas Number : 1099621-62-1
  • Molecular Weight : 261.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65943

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65943
Availability details for
Quantity
  • Availability
  • Add to Cart