ENA300790907 Enamine

1-amino-3-(4-iodophenoxy)propan-2-ol

  • MDL number MFCD11182824
  • Cas Number : 857347-04-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65973

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65973
Availability details for
Quantity
  • Availability
  • Add to Cart