ENA300790910 Enamine

3-[(5-cyclopropyl-1,3,4-oxadiazol-2-yl)carbamoyl]propanoic acid

  • MDL number MFCD12112379
  • Cas Number : 1156424-49-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65979

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65979
Availability details for
Quantity
  • Availability
  • Add to Cart