ENA300790911 Enamine

ethyl 2-(furan-2-yl)-4-methyl-6-oxo-1,6-dihydropyrimidine-5-carboxylate

  • MDL number MFCD09403622
  • Cas Number : 941190-95-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-34791

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-34791
Availability details for
Quantity
  • Availability
  • Add to Cart