ENA300790914 Enamine

2-tert-butyl-4-methyl-6-oxo-1,6-dihydropyrimidine-5-carboxylic acid

  • MDL number MFCD09802121
  • Cas Number : 949792-16-9
  • Molecular Weight : 210.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-34846

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-34846
Availability details for
Quantity
  • Availability
  • Add to Cart