ENA300790917 Enamine

4-amino-2-methoxy-N-methylbenzene-1-sulfonamide

  • MDL number MFCD11621463
  • Cas Number : 1094904-92-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65987

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65987
Availability details for
Quantity
  • Availability
  • Add to Cart