ENA300790922 Enamine

quinoline-4-thiol

  • MDL number MFCD00956872
  • Cas Number : 51812-96-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 159...161
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65999

Safety Information

msds https://www.enaminestore.com/catalog/EN300-65999
Availability details for
Quantity
  • Availability
  • Add to Cart