ENA300790923 Enamine

4-oxo-1,4-dihydroquinoline-3-carbonitrile

  • MDL number MFCD00957040
  • Cas Number : 71083-59-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66000

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-66000
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart