ENA300790924 Enamine

2-methyl-6-(piperidine-4-carbonyl)-3,4-dihydro-2H-1,4-benzoxazin-3-one hydrochloride

  • MDL number MFCD16817437
  • Cas Number : 1241340-57-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66003

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66003
Availability details for
Quantity
  • Availability
  • Add to Cart