ENA300790930 Enamine

4-(4-hydroxypiperidine-1-carbonyl)-1-methyl-1,2-dihydropyridin-2-one

  • MDL number MFCD12416555
  • Cas Number : 1155607-95-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66010

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66010
Availability details for
Quantity
  • Availability
  • Add to Cart