ENA300790938 Enamine

4-cyclopropyl-5-[(4-fluorophenyl)methyl]-1,3-thiazol-2-amine

  • MDL number MFCD16817440
  • Cas Number : 1251922-72-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds https://www.enaminestore.com/catalog/EN300-66032
Availability details for
Quantity
  • Availability
  • Add to Cart