ENA300790944 Enamine

2-(ethanesulfonyl)-1,2,3,4-tetrahydroisoquinolin-5-amine

  • MDL number MFCD12821605
  • Cas Number : 1182836-84-5
  • Molecular Weight : 240.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(ethylsulfonyl)-1,2,3,4-tetrahydro-5-isoquinolinamine
Inchi Code 1S/C11H16N2O2S/c1-2-16(14,15)13-7-6-10-9(8-13)4-3-5-11(10)12/h3-5H,2,6-8,12H2,1H3
Inchi Key BYQPVXVPDNRJLV-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-66041

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66041
Availability details for
Quantity
  • Availability
  • Add to Cart