ENA300790946 Enamine

1-(3,4-dimethylphenyl)ethan-1-amine

  • MDL number MFCD02177054
  • Cas Number : 91251-29-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-35203

Safety Information

msds https://www.enaminestore.com/catalog/EN300-35203
Availability details for
Quantity
  • Availability
  • Add to Cart