ENA300790951 Enamine

2-(furan-2-yl)-4-methyl-6-oxo-1,6-dihydropyrimidine-5-carboxylic acid

  • MDL number MFCD09403765
  • Cas Number : 941166-92-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-35452

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-35452
Availability details for
Quantity
  • Availability
  • Add to Cart