ENA300790958 Enamine

N-(2-isothiocyanatoethyl)methanesulfonamide

  • MDL number MFCD12135310
  • Cas Number : 1155623-89-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66060

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66060
Availability details for
Quantity
  • Availability
  • Add to Cart