ENA300790959 Enamine

[4-(propan-2-yloxy)phenyl]thiourea

  • MDL number MFCD11202364
  • Cas Number : 420120-01-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-66061

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66061
Availability details for
Quantity
  • Availability
  • Add to Cart