ENA300790960 Enamine

3-amino-1-[2-(propan-2-yl)phenyl]thiourea

  • MDL number MFCD11188561
  • Cas Number : 1039924-20-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66062

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-66062
pictograms GHS06
Availability details for
Quantity
  • Availability
  • Add to Cart