ENA300790962 Enamine

5-(2,5-dimethylfuran-3-yl)-1,3,4-oxadiazol-2-amine

  • MDL number MFCD09041623
  • Cas Number : 926218-66-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66064

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66064
Availability details for
Quantity
  • Availability
  • Add to Cart