ENA300790968 Enamine

propan-2-yl 3-amino-4-chlorobenzoate

  • MDL number MFCD08685817
  • Cas Number : 343773-02-4
  • Molecular Weight : 213.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66071

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66071
Availability details for
Quantity
  • Availability
  • Add to Cart