ENA300790969 Enamine

3-(3,3-dimethylbutanoyl)-1,3-thiazolidine-4-carboxylic acid

  • MDL number MFCD09945447
  • Cas Number : 1103859-83-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66072

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66072
Availability details for
Quantity
  • Availability
  • Add to Cart