ENA300790975 Enamine

1-phenyl-1H-1,2,3,4-tetrazol-5-amine

  • MDL number MFCD00083048
  • Cas Number : 5467-78-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 165-168
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66080

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66080
Availability details for
Quantity
  • Availability
  • Add to Cart