ENA300790984 Enamine

ethyl 3-amino-3-(4-bromophenyl)propanoate hydrochloride

  • MDL number MFCD02663241
  • Cas Number : 502841-92-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66102

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66102
Availability details for
Quantity
  • Availability
  • Add to Cart