ENA300790990 Enamine

2-hydroxy-3-phenylbutanoic acid

  • MDL number MFCD14622119
  • Cas Number : 91061-47-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-66113

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66113
Availability details for
Quantity
  • Availability
  • Add to Cart