ENA300790991 Enamine

3-(4,5,6,7-tetrahydro-1,3-benzothiazol-2-yl)propan-1-amine

  • MDL number MFCD09891579
  • Cas Number : 933734-44-2
  • Molecular Weight : 196.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-66114

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66114
Availability details for
Quantity
  • Availability
  • Add to Cart