ENA300790995 Enamine

4-methyl-2-phenylpiperazine-1-sulfonamide

  • MDL number MFCD11205218
  • Cas Number : 1094315-38-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-66122

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66122
Availability details for
Quantity
  • Availability
  • Add to Cart