ENA300790998 Enamine

2-(4-chlorophenyl)propan-2-ol

  • MDL number MFCD11870102
  • Cas Number : 1989-25-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 41-45
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66125

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66125
Availability details for
Quantity
  • Availability
  • Add to Cart