ENA300790999 Enamine

(1R)-1-(thiophen-2-yl)ethan-1-ol

  • MDL number MFCD06797334
  • Cas Number : 86527-10-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-66136

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66136
Availability details for
Quantity
  • Availability
  • Add to Cart