ENA300791008 Enamine

2-methyl-1,2,3,4-tetrahydroquinoline-1-sulfonamide

  • MDL number MFCD11205220
  • Cas Number : 1094222-89-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66148

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66148
Availability details for
Quantity
  • Availability
  • Add to Cart