ENA300791009 Enamine

2-methyl-1,4-oxazepane

  • MDL number MFCD10035564
  • Cas Number : 1018614-41-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66149

Safety Information

msds https://www.enaminestore.com/catalog/EN300-66149
Availability details for
Quantity
  • Availability
  • Add to Cart