ENA300791017 Enamine

4-[(thiomorpholin-4-yl)methyl]aniline

  • MDL number MFCD11172085
  • Cas Number : 210686-44-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-66157

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66157
Availability details for
Quantity
  • Availability
  • Add to Cart