ENA300791019 Enamine

2-({[(pyridin-3-yl)methyl]carbamoyl}methoxy)acetic acid

  • MDL number MFCD01029152
  • Cas Number : 312924-82-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-66159

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66159
Availability details for
Quantity
  • Availability
  • Add to Cart