ENA300791022 Enamine

2-[(pyridin-4-yl)amino]ethan-1-ol

  • MDL number MFCD08437502
  • Cas Number : 192130-06-6
  • Molecular Weight : 138.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-66163

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66163
Availability details for
Quantity
  • Availability
  • Add to Cart