ENA300791023 Enamine

N-(piperidin-4-yl)cyclobutanecarboxamide

  • MDL number MFCD01763801
  • Cas Number : 953899-02-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66164

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66164
Availability details for
Quantity
  • Availability
  • Add to Cart