ENA300791024 Enamine

5-[(4-methanesulfonylphenoxy)methyl]furan-2-carboxylic acid

  • MDL number MFCD13805638
  • Cas Number : 1157777-00-8
  • Molecular Weight : 296.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66165

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66165
Availability details for
Quantity
  • Availability
  • Add to Cart